Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023050 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3050 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-3050 Supplier MilliporeSigma Supelco Supplier No. RTC00009220ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -90°C (literature)
CAS 71-36-3
Density 0.81 g/mL (at 25°C (literature))
Quantity 20 mL
Flash Point 35°C (Not Applicable)
Refractive Index n20/D 1.399 (literature)
Molecular Formula C4H10O
Linear Formula CH3(CH2)3 OH
MDL Number MFCD00002964
Synonym 1-Butanol; n-Butanol; Butyl alcohol
Boiling Point 116°C to 118°C (literature)
Formula Weight 74.12
Grade Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.