Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice

Catalog No. 6000088 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-000-88 2.5 L
60-000-90 4 x 2.5 L
60-048-234 4 x 4 L
60-000-87 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-000-88 Supplier Solstice Supplier No. 348672.5L
Add to Cart
Add to Cart

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Density 0.81g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
MDL Number MFCD00002964
Beilstein 969148
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
Boiling Point 116°C to 118°C
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV™ Plus
Chemical Name or Material 1-Butanol, for HPLC
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H302-H315-H318-H335-H336
EINECSNumber 200-751-6
RTECSNumber EO1400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.