Learn More
1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice
Shop All Solstice Research Chemicals Products

Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Melting Point | -90°C (literature) |
| Absorbance | max. 0.10 at 240 nm, max. 0.10 at 235 nm, max. 0.50 at 225 nm, max. 0.70 at 220 nm, max. 1.0 at 210 nm |
| CAS Max % | 100.0000% |
| Density | 0.81 g/mL at 25 °C(lit.) |
| Quantity | 4 x 4 L |
| Flash Point | 35°C |
| Packaging | Glass Bottle |
| Linear Formula | C6H5CH3 |
| MDL Number | MFCD00008512 |
| UN Number | UN1120 |
| Show More |
Safety and Handling
| ShelfLife | 720 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butanols |
| EINECSNumber | 200-751-6 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.