Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017052 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7052 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7052 Supplier MilliporeSigma Supelco Supplier No. PHR12093X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 1-Butanol is a potential fuel substitute to gasoline. It is also a feedstock used to produce chemicals. It is produced via two pathways, the synthetic 2-ketoacid pathway and CoA-dependent pathway. It can also be produced by fermentation of biomass-derived carbohydrates. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -90°C
CAS 71-36-3
Density 0.81 g/mL (at 25°C)
Quantity 3 x 1.2 mL
Flash Point 35°C - Pensky-Martens closed cup
Ignition Point 649°F
Physical Form Neat
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula CH3(CH2)3 OH
MDL Number MFCD00002964
Synonym n-Butanol; Butyl alcohol
Boiling Point 116°C to 118°C
Formula Weight 74.12
Grade Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.