Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Butanol suitable for HPL
Catalog No. 502899312
Encompass
1-Butanol is a linear alcohol. Its biosynthesis from glucose using a bioengineered Escherichia coli strain has been reported. Its reaction with diisocyanates in the presence of triethylamine (catalyst) has been investigated by volumetric method. Kinetic studies suggest that the reaction proceeds via competitive consecutive second order. Its dehydration reaction has been employed as a standard reaction to evaluate the acidic and basic characteristics of various silica-aluminas.
Catalog No. 50-289-9312 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348672L
1-Butanol is a linear alcohol. Its biosynthesis from glucose using a bioengineered Escherichia coli strain has been reported. Its reaction with diisocyanates in the presence of triethylamine (catalyst) has been investigated by volumetric method. Kinetic studies suggest that the reaction proceeds via competitive consecutive second order. Its dehydration reaction has been employed as a standard reaction to evaluate the acidic and basic characteristics of various silica-aluminas.