Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-1-methylpiperidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. B48445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48445G 5 g
B484425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48445G Supplier TCI America Supplier No. B48445G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623580-02-9
Molecular Formula C12H22F6N2O4S2
Molecular Weight (g/mol) 436.43
MDL Number MFCD14156017
InChI Key ZDMWZUAOSLBMEY-UHFFFAOYSA-N
PubChem CID 12184311
IUPAC Name 1-butyl-1-methylpiperidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
SMILES CCCC[N+]1(C)CCCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 436.43
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-methylpiperidinium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.