Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-1-Methylpiperidinium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 7130745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-307-45G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-307-45G Supplier Sigma Aldrich Supplier No. 7130745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide is an ionic liquid that can be used:As an electrolyte in ionic liquid dual ion battery and dual-graphite battery.As a model ionic liquid in the study of ionic effect on the desulfurization of fuels.In the study of the activity coefficient of different solutes at infinite dilution.

Specifications

CAS 402846-78-0
Melting Point 37°C; 37°C
Density 1.198 g/mL at 20°C
Molecular Formula C9H17BF4N2
Linear Formula C9H17BF4N2
MDL Number MFCD03427618
Quantity 50 g
Molecular Weight (g/mol) 240.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.