Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-1-Methylpyrrolidinium Bis(Trifluoromethylsulfonyl)Imide

Catalog No. 388945GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-894-5GF 5 g
38-894-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-894-5GF Supplier Sigma Aldrich Supplier No. 388945GF
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-1-Methylpyrrolidinium Bis(Trifluoromethylsulfonyl)Imide

Specifications

CAS 223437-11-4
Melting Point -18°C
Molecular Formula C11H20F6N2O4S2
Linear Formula C11H20F6N2O4S2
MDL Number MFCD07784447
Quantity 5 g
Synonym BMP-BTI
Molecular Weight (g/mol) 422.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.