Learn More
Medchemexpress LLC 1-butyl-1-methylpyrrolidinium chloride | 479500-35-1 | MFCD08458922 | ≥98.0% | 177.72 | C9H20ClN | 25 G
SDP
Catalog No. 5000302317
Encompass_Preferred
1-Butyl-1-methylpyrrolidinium chloride is an ionic liquid (a quaternary ammonium chloride) commonly used as a solvent and reaction medium in organic synthesis, electrochemistry, and materials research. Supplied for research use with accompanying technical and safety documentation, it offers a defined composition and high purity suitable for laboratory applications.
  • High purity (≥98.0%).
  • Ionic liquid solvent suitable for synthesis and electrochemical applications.
  • Solid at room temperature with good thermal stability.
  • Available in multiple pack sizes for laboratory use.
  • Comes with technical datasheet and safety data sheet for handling guidance.
Catalog No. 50-003-02317 Supplier Medchemexpress LLC Supplier No. 25GHYW016638
Add to Cart
Add to Cart
1-Butyl-1-methylpyrrolidinium chloride is an ionic liquid (a quaternary ammonium chloride) commonly used as a solvent and reaction medium in organic synthesis, electrochemistry, and materials research. Supplied for research use with accompanying technical and safety documentation, it offers a defined composition and high purity suitable for laboratory applications.
  • High purity (≥98.0%).
  • Ionic liquid solvent suitable for synthesis and electrochemical applications.
  • Solid at room temperature with good thermal stability.
  • Available in multiple pack sizes for laboratory use.
  • Comes with technical datasheet and safety data sheet for handling guidance.