Learn More
Medchemexpress LLC 1-butyl-1-methylpyrrolidinium chloride | 479500-35-1 | MFCD08458922 | >98.0% | 177.72 g·mol⁻¹ | C9H20ClN | 5 G
SDP
Catalog No. 5000301129
Encompass_Preferred
1-Butyl-1-methylpyrrolidinium chloride is a quaternary ammonium salt provided as a white to off-white solid for use as a reagent and as an ionic-liquid component in laboratory research. It is produced at high purity and characterized by its defined molecular weight and formula, making it suitable for synthesis and analytical workflows.
  • High purity ≥98.0% suitable for laboratory use.
  • Solid form, white to off-white.
  • Molecular weight 177.72 g·mol⁻¹; formula C9H20ClN.
  • Available in small-scale packaging, including 5 g.
  • Used as a reagent and ionic-liquid component in synthesis and analysis.
Catalog No. 50-003-01129 Supplier Medchemexpress LLC Supplier No. HYW0166385G
1-Butyl-1-methylpyrrolidinium chloride is a quaternary ammonium salt provided as a white to off-white solid for use as a reagent and as an ionic-liquid component in laboratory research. It is produced at high purity and characterized by its defined molecular weight and formula, making it suitable for synthesis and analytical workflows.
  • High purity ≥98.0% suitable for laboratory use.
  • Solid form, white to off-white.
  • Molecular weight 177.72 g·mol⁻¹; formula C9H20ClN.
  • Available in small-scale packaging, including 5 g.
  • Used as a reagent and ionic-liquid component in synthesis and analysis.