Learn More
Medchemexpress LLC 1-butyl-1-methylpyrrolidinium chloride | 479500-35-1 | MFCD08458922 | 98.0% | 177.72 g·mol⁻¹ | C9H20ClN | 50 G
SDP
Catalog No. 5000300799
Encompass_Preferred
1-Butyl-1-methylpyrrolidinium chloride is a quaternary pyrrolidinium chloride salt used as an ionic liquid and laboratory reagent. It is employed in research requiring stable, low-volatility ionic liquids, and as a reagent or intermediate in chemical synthesis.
  • Quaternary ammonium ionic liquid suitable for research use.
  • High purity (~98%) for reproducible results.
  • Low volatility and good thermal stability.
  • Soluble in polar solvents, compatible with many reaction media.
  • Available in small lab-scale pack sizes for bench work.
Catalog No. 50-003-00799 Supplier Medchemexpress LLC Supplier No. HYW01663850G
1-Butyl-1-methylpyrrolidinium chloride is a quaternary pyrrolidinium chloride salt used as an ionic liquid and laboratory reagent. It is employed in research requiring stable, low-volatility ionic liquids, and as a reagent or intermediate in chemical synthesis.
  • Quaternary ammonium ionic liquid suitable for research use.
  • High purity (~98%) for reproducible results.
  • Low volatility and good thermal stability.
  • Soluble in polar solvents, compatible with many reaction media.
  • Available in small lab-scale pack sizes for bench work.