Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-1-Methylpyrrolidinium Dicyanamide

Catalog No. 5089350GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-893-50GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 50-893-50GF Supplier Sigma Aldrich Supplier No. 5089350GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-1-methylpyrrolidinium dicyanamide is a hypergolic, air- and water-stable pyrrolidinium-based ionic liquid. It shows a wide potential range, making it a promising candidate in electrochemical applications. [BMPYR][DCA] also has high extraction capacity for dibenzothiophene fro mLiquid fuels.

Specifications

CAS 68716-47-2
Melting Point 246°C to 249°C
Molecular Formula C6H5BCl2O2
Linear Formula Cl2C6H3B(OH)2
Quantity 5 g
MDL Number MFCD00013930
Synonym 2,4-Dichlorobenzeneboronic acid
Molecular Weight (g/mol) 190.82 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.