Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2,3-dimethylimidazolium Polyethylene Glycol Hexadecyl Ether Sulfate coated Lipase, TCI America™
SDP

Catalog No. B30281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30281G Supplier TCI America Supplier No. B30281G
Add to Cart
Add to Cart

Chemical Identifiers

Synonym Lipase coated with 1-Butyl-2,3-dimethylimidazolium Polyethylene Glycol Hexadecyl Ether Sulfate

Specifications

Color Yellow
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium Polyethylene Glycol Hexadecyl Ether Sulfate coated Lipase
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.