Learn More
Strem, An Ascensus Company CAS# 174501-65-6. 5g. 1-Butyl-3-methylimidazolium tetrafluoroborate, 98% [BMIM] [BF4]. MFCD03095449
SDP
Catalog No. 5090108897
Encompass
CAS# 174501-65-6. 5g. 1-Butyl-3-methylimidazolium tetrafluoroborate, 98% [BMIM] [BF4]. MFCD03095449. Molecular Weight: 226.03. Molecular Formula: [C8H15N2]+BF4ae. Color/form: colorless to yellow liq Strem# 07-0170. http://www.strem.com/catalog/v/07-0170/
Catalog No. 50-901-08897 Supplier Strem, An Ascensus Company Supplier No. 0701705G
CAS# 174501-65-6. 5g. 1-Butyl-3-methylimidazolium tetrafluoroborate, 98% [BMIM] [BF4]. MFCD03095449. Molecular Weight: 226.03. Molecular Formula: [C8H15N2]+BF4ae. Color/form: colorless to yellow liq Strem# 07-0170. http://www.strem.com/catalog/v/07-0170/