Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Acetate

Catalog No. 303675G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-367-5G 5 g
30-367-50G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-367-5G Supplier Sigma Aldrich Supplier No. 303675G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium acetate is a room temperature ionic liquid. It is reported to irreversibly adsorb gaseous CO2, therefore can be a useful means for the separation of greenhouse gas. CO2 has been reported to readily dissolve in 1-butyl-3-methylimidazolium-acetate. It also forms reversible molecular complex with CO2 in the ratio of 1 (CO2) : 2 (1-butyl-3-methylimidazolium-acetate).1-Butyl-3-methylimidazolium acetate may be used to effectively capture CO2 from post-combustion flue gas. It may be used in the preparation of 1-butyl-3-methylimidazolium-2-carboxylate via carboxylation with CO2.

Specifications

CAS 284049-75-8
Molecular Formula C10H18N2O2
Linear Formula C10H18N2O2
MDL Number MFCD06798175
Quantity 5 g
Molecular Weight (g/mol) 198.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.