Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. B24775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24775G 5 g
B247725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24775G Supplier TCI America Supplier No. B24775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 174899-83-3
Molecular Formula C10H15F6N3O4S2
Molecular Weight (g/mol) 419.357
MDL Number MFCD05664714
InChI Key INDFXCHYORWHLQ-UHFFFAOYSA-N
PubChem CID 11258643
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point -2°C
Color Yellow
Quantity 5 g
Formula Weight 419.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.