Learn More
Medchemexpress LLC 1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide | 174899-83-3 | HY-W010937 | 419.36 | 25 G
SDP
Catalog No. 5000451794
Encompass_Preferred
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is an HIV-1 integrase inhibitor that effectively inhibits both the 3'-processing (3'-P) and strand transfer (ST) steps of the integration reaction. It can be utilized in HIV-1 research.
  • Functions as an HIV-1 integrase inhibitor
  • Effectively inhibits 3'-processing (3'-P) steps
  • Effectively inhibits strand transfer (ST) steps
  • Suitable for HIV-1 research
Catalog No. 50-004-51794 Supplier Medchemexpress LLC Supplier No. 25GHYW010937
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is an HIV-1 integrase inhibitor that effectively inhibits both the 3'-processing (3'-P) and strand transfer (ST) steps of the integration reaction. It can be utilized in HIV-1 research.
  • Functions as an HIV-1 integrase inhibitor
  • Effectively inhibits 3'-processing (3'-P) steps
  • Effectively inhibits strand transfer (ST) steps
  • Suitable for HIV-1 research