Learn More
Medchemexpress LLC 1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide | 174899-83-3 | MFCD05664714 | 99.9% | 419.36 | 50 G
SDP
Catalog No. 5000366281
Encompass_Preferred
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide (compound 8) is an HIV-1 integrase inhibitor that effectively inhibits both the 3'-processing (3'-P) and strand transfer (ST) steps of the integration reaction. It can be utilized in HIV-1 research.
  • Effectively inhibits HIV-1 integrase
  • Inhibits 3'-processing step
  • Inhibits strand transfer step
  • Suitable for HIV-1 research
Catalog No. 50-003-66281 Supplier Medchemexpress LLC Supplier No. HYW01093750G
Add to Cart
Add to Cart
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide (compound 8) is an HIV-1 integrase inhibitor that effectively inhibits both the 3'-processing (3'-P) and strand transfer (ST) steps of the integration reaction. It can be utilized in HIV-1 research.
  • Effectively inhibits HIV-1 integrase
  • Inhibits 3'-processing step
  • Inhibits strand transfer step
  • Suitable for HIV-1 research