Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

Catalog No. 90080225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-080-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-080-225G Supplier Sigma Aldrich Supplier No. 90080225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.Ionic liquids (ILs) are molten salts with melting points lower than 100°C. They usually consist of pair of organic cation and anion. ILs exhibit unique properties such as non-volatility, high thermal stability, and high ionic conductivity and find applications as electrolytes in lithium/sodium ion batteries and dye-sensitized solar cells. They are also used as media for synthesis of conducting polymers and intercalation electrode materials.

Specifications

CAS 174899-83-3
Molecular Formula C10H15F6N3O4S2
Linear Formula C10H15F6N3O4S2
Quantity 25 g
MDL Number MFCD05664714
Synonym 1-Butyl-3-methylimidazolium bis(trifluoromethanesulfonimide) salt; 1-Butyl-3-methylimidazolium bis(trifluoromethanesulfonyl)amide; 1-Butyl-3-methylimidazolium bis(triflyl)imide; BMIIm; BMIM TFSI
Molecular Weight (g/mol) 419.36
Percent Purity ≥99%
Biological Activity Nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.