Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Bromide

Catalog No. 9513750GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-137-50GF 5 g
95-137-5GF 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 95-137-50GF Supplier Sigma Aldrich Supplier No. 9513750GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium bromide is a neutral ionic liquid.BMIMBr undergoes gelation with gelatin to form ion jelly, a quasi-solid material for use in chemoresistive gas sensors. It can be used as a solvent for the preparation of 1,2,4,5-substituted imidazoles.Ionic liquid used in a Heck reaction

Specifications

CAS 85100-77-2
Molecular Formula C8H15BrN2
Linear Formula C8H15BrN2
MDL Number MFCD03427611
Quantity 5 g
Synonym BMIMBr
Molecular Weight (g/mol) 219.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.