Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
79917-90-1
C8H15ClN2
174.67
MFCD03095425
FHDQNOXQSTVAIC-UHFFFAOYSA-M
1-butyl-3-methylimidazolium chloride,3-butyl-1-methyl-1h-imidazol-3-ium chloride,1-n-butyl-3-methylimidazolium chloride,bmimcl,bmim cl,c4mim chloride,dsstox_cid_11461,dsstox_rid_78878,dsstox_gsid_31461
2734161
1-butyl-3-methylimidazol-3-ium;chloride
CCCCN1C=C[N+](=C1)C.[Cl-]
79917-90-1
C8H15ClN2
174.67
MFCD03095425
FHDQNOXQSTVAIC-UHFFFAOYSA-M
1-butyl-3-methylimidazolium chloride,3-butyl-1-methyl-1h-imidazol-3-ium chloride,1-n-butyl-3-methylimidazolium chloride,bmimcl,bmim cl,c4mim chloride,dsstox_cid_11461,dsstox_rid_78878,dsstox_gsid_31461
2734161
1-butyl-3-methylimidazol-3-ium;chloride
CCCCN1C=C[N+](=C1)C.[Cl-]
Ionic liquid
Color Cream to Yellow
Packaging Glass bottle
Viscosity 170.1 mPa.s (80°C)
Melting Point 73.0°C
Quantity 25g
Infrared Spectrum Authentic
Water 1% Max. (K.F.)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Merck Index 15, 1584
Solubility Information Solubility in water: soluble
Formula Weight 174.67
Physical Form Mass
Percent Purity 98+%
Chemical Name or Material 1-Butyl-3-methylimidazolium chloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a

GHS Signal Word: Danger

TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.