Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Dicyanamide

Catalog No. 552205GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-220-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 55-220-5GF Supplier Sigma Aldrich Supplier No. 552205GF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-Methylimidazolium Dicyanamide

Specifications

CAS 17791-51-4
Melting Point 113°C to 116°C
Molecular Formula C17H27N3O6
Linear Formula C17H27N3O6
MDL Number MFCD03094049
Quantity 5 g
Synonym [Methyl(S)-3[1-(1,1-dimethylethoxycarbonyl)-1-H-imidazol-4-yl]-2-[(1,1-dimethylethoxycarbonyl)amino]propionate]
Molecular Weight (g/mol) 369.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.