Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Hexafluorophosphate

Catalog No. 709565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-956-5G 5 g
70-956-250G 250 g
70-956-50G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 70-956-5G Supplier Sigma Aldrich Supplier No. 709565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium hexafluorophosphate is an imidazolium-based, hydrophobic, room temperature ionic liquid (RTIL). It can be prepared by reacting 1-methylimidazole with chlorobutane. Gaseous hydrofluorocarbons (HFCs) such as fluoromethane, fluoroethane and 1,1,2,2-tetrafluoroethane are soluble in BMIMPF6.1-Butyl-3-methylimidazolium hexafluorophosphate is an ionic liquid employed in many environmentally friendly reactions. It can also be used as a medium for reactions such as: Ring-closing metathesis of diene and enyne substrates in the presence of a novel recyclable ruthenium carbene complex.Nickel(II)acetylacetonate catalyzed oxidation of aromatic aldehydes to the corresponding acids using dioxygen as the oxidant.Lipase-catalyzed enantioselective acylation of allylic alcohols.Allylation of aldehydes using tetraallylstannane to yield homoallylic alcohols.

Specifications

CAS 174501-64-5
Density 1.38 g/mL at 20°C
Molecular Formula C8H15F6N2P
Refractive Index n20/D 1.411
Linear Formula C8H15F6N2P
MDL Number MFCD03093295
Quantity 5 g
Synonym BMIMPF6
Molecular Weight (g/mol) 284.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.