Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. B232025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B232025G 25 g
B23205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B232025G Supplier TCI America Supplier No. B232025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 284.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.