Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Hydrogen Sulfate

Catalog No. 574571KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-457-1KGF 1 kg
57-457-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 57-457-1KGF Supplier Sigma Aldrich Supplier No. 574571KGF
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.[BMIM][HSO4] can be used as an additive to accelerate current efficiency, lower power consumption, and enhance the surface morphology during the electrodeposition of zinc from acidic sulfate solutions. The combination of sodium nitrate and [BMIM][HSO4] can be used as a nitrating agent for the selective nitration of phenols to afford its corresponding nitro derivatives. [BMIM][HSO4] can also be used as catalyst for the preparation of 1,8-dioxo-octahydroxanthenes.

Specifications

CAS 262297-13-2
Molecular Formula C8H16N2O4S
Linear Formula C8H16N2O4S
MDL Number MFCD06798197
Quantity 1 kg
Synonym [BMIM][HSO4]
Molecular Weight (g/mol) 236.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.