Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Iodide

Catalog No. 7130665G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-306-65G 5 g
71-306-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 71-306-65G Supplier Sigma Aldrich Supplier No. 7130665G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium iodide is a room temperature ionic liquid (RTIL).BMImI can be used as a non-volatile electrolyte in dye-sensitized photochemical cells. It can also be used as a stabilizer/promotor/reaction medium for the transition metal catalyzed biphasic oxidative cyclocarbonylation of +a-aminoalcohols and 2-aminophenol to form the corresponding 2-oxazolidinones.

Specifications

CAS 65039-05-6
Molecular Formula C8H15IN2
Linear Formula C8H15IN2
MDL Number MFCD08276307
Quantity 5 g
Synonym BMIMI
Molecular Weight (g/mol) 266.12 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.