Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Octyl Sulfate

Catalog No. 750595GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-059-5GF 5 g
75-059-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 75-059-5GF Supplier Sigma Aldrich Supplier No. 750595GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium octyl sulfate is a halogen-free ionic liquid. It behaves as a surfactant when the critical micelle concentration (CMC) is above 0.031M.1-Butyl-3-methylimidazolium octyl sulfate can be used as a solvent during the hydroformylation of 1-octene to form nonanals in the presence of a rhodium catalyst.Increases the yield and enzyme stability of +a-galactosidase in enzyme-catalyzed syntheses

Specifications

CAS 445473-58-5
Molecular Formula C16H32N2O4S
Linear Formula C16H32N2O4S
MDL Number MFCD03788914
Quantity 5 g
Synonym BMIM OSU
Molecular Weight (g/mol) 348.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.