Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Tetrachloroaluminate

Catalog No. 55292100GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-292-100GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger-Danger
Catalog No. 55-292-100GF Supplier Sigma Aldrich Supplier No. 55292100GF
Only null left
Add to Cart
Add to Cart
Danger-Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium tetrachloroaluminate is an ionic liquid that can be prepared by reacting 1-butyl-3-methylimidazolium chloride with aluminum chloride hexahydrate.1-Butyl-3-methylimidazolium tetrachloroaluminate can be used as an efficient catalyst in tetrahydropyranylation of alcohols, and detetrahydropyranylation of THP (tetrahydropyranyl) protected ethers. It can also be used as: A catalyst to synthesize linear alkylbenzenes by alkylation of benzene with 1-dodecene.A solvent in extractive desulfurization of oil products.

Specifications

CAS 94964-58-6
Melting Point 65°C to 69°C
Molecular Formula C14H23NO4
Linear Formula C14H23NO4
MDL Number MFCD03093958
Quantity 1 g
Molecular Weight (g/mol) 269.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.