Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Tetrafluoroborate

Catalog No. 915085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
5 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-508-5G 250 g
91-508-50G 5 g
91-508-250G 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 91-508-5G Supplier Sigma Aldrich Supplier No. 915085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium tetrafluoroborate may be used as an extractant for the removal of dibenzothiophene (DBS) fro mLiquid fuels via extractive desulfurization.Ionic liquid used in many reactions, e.g. hydrogenations; asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase; Suzuki cross-coupling at ambient temperature

Specifications

CAS 174501-65-6
Density 1.21 g/mL at 20°C
Molecular Formula C8H15BF4N2
Linear Formula C8H15BF4N2
MDL Number MFCD03095449
Quantity 250 g
Synonym BMIMBF4
Molecular Weight (g/mol) 226.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.