Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Tetrafluoroborate

Catalog No. 7117481KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-174-81KG 1 kg
71-174-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 71-174-81KG Supplier Sigma Aldrich Supplier No. 7117481KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium tetrafluoroborate is a hydrophilic room temperature ionic liquid (RTIL).1-Butyl-3-methylimidazolium tetrafluoroborate ([BMIM]BF4) can be used: As a reaction medium for the preparation NH4TiOF3 mesocrystals, which are converted into TiO2 based nanostructures As a working fluid along with 2,2,2-trifluoroethanol in absorption heat pumps or chillers As an electrolyte in lithium-ion batteries and double layer capacitors.

Specifications

CAS 174501-65-6
Density 1.21 g/mL at 20°C
Molecular Formula C8H15BF4N2
Linear Formula C8H15BF4N2
MDL Number MFCD03095449
Quantity 1 kg
Synonym BMIMBF4
Molecular Weight (g/mol) 226.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.