Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%

Catalog No. AC354211000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC354211000 100 g Glass bottle
AC354210050 5 g Glass bottle
AC354210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC354211000 Supplier Thermo Scientific Chemicals Supplier No. 354211000
  • $423.00
    $338.65 / Each of 1

    Save $84.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
PubChem CID 2734178
IUPAC Name 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -71.0°C
Color Orange-Yellow
Density 1.2077g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 1584
Viscosity 118.3 mPa.s (25°C)
Formula Weight 226.02
Percent Purity ≥98%
Physical Form Viscous Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium tetrafluoroborate
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.