Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Thiocyanate

Catalog No. 42254100GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-254-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-254-100GF Supplier Sigma Aldrich Supplier No. 42254100GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium thiocyanate ([BMIM][SCN]) ionic liquid can be used:As an alternative to conventional entrainers in the extractive distillation of cyclohexane and benzene.In polyethylene oxide-sodium perchlorate electrolyte to increase its ionic conductivity at room temperature.[BMIM][SCN] show higher degree of solubility of lithium salts such as lithium perchlorate (LiClO4) and N-lithiotrifluoromethanesulfonimide(LiTFSI) when compared to other room temperature ionic liquids due to the presence of polarizable anion (-SCN).

Specifications

CAS 69500-64-7
Density 0.933 g/mL at 25°C
Boiling Point 65°C (11 mmHg, lit.)
Molecular Formula C5H12N2
Linear Formula C5H12N2
Quantity 100 g
MDL Number MFCD00191745
Molecular Weight (g/mol) 100.16 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.