Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Tribromide 98.0+%, TCI America™
SDP

Catalog No. B35965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35965G Supplier TCI America Supplier No. B35965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 820965-08-0
Molecular Formula C8H15Br3N2
Molecular Weight (g/mol) 378.94
MDL Number MFCD22199811
InChI Key RHSQXXVEZYTZHB-UHFFFAOYSA-K
PubChem CID 91983981
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium tribromide
SMILES [Br-].[Br-].[Br-].CCCCN1C=C[N+](C)=C1

Specifications

Melting Point 8°C
Quantity 5 g
Formula Weight 378.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Tribromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.