Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Trifluoro(trifluoromethyl)borate 98.0+%, TCI America™
SDP

Catalog No. B35425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35425G Supplier TCI America Supplier No. B35425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 741677-68-9
Molecular Formula C9H15BF6N2
Molecular Weight (g/mol) 276.033
InChI Key PTBVLMHYOBXNCB-UHFFFAOYSA-N
PubChem CID 44630030
IUPAC Name 1-butyl-3-methylimidazol-3-ium;trifluoro(trifluoromethyl)boranuide
SMILES [B-](C(F)(F)F)(F)(F)F.CCCCN1C=C[N+](=C1)C

Specifications

Quantity 5 g
Formula Weight 276.03
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Trifluoro(trifluoromethyl)borate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.