Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Trifluoromethanesulfonate

Catalog No. 711772100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-177-2100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-177-2100G Supplier Sigma Aldrich Supplier No. 711772100G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium trifluoromethanesulfonate [bmim][OTf] is a versatile ionic liquid commonly used in synthetic chemistry.[bmim][OTf] can be used as: A glycosylation promoter in the synthesis of oligosaccharides from thiophenyl and trichloroacetimidate glycoside donorsA solvent for the extraction of sulfur and nitrogen containing aromatic organic compounds from aliphatic hydrocarbons.A reagent in the preparation of ion-conductive polymeric membranes A dopant in the synthesis of poly (ethyl methacrylate) based polymer electrolytes to increase the ionic conductivity.A reaction medium to synthesize aryl ketones by Friedel-Crafts benzoylation of aryl compounds by using bismuth triflate as a catalyst.

Specifications

CAS 56883-39-7
Melting Point >300°C
Molecular Formula C44H44N2P2
Refractive Index n20/D 1.434
Linear Formula C44H44N2P2
Quantity 250 mg
MDL Number MFCD16618374
Synonym BMIM Otf
Molecular Weight (g/mol) 662.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.