Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Trifluoromethanesulfonate

Catalog No. 7642025GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-420-25GF 5 g
76-420-5GF 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-420-25GF Supplier Sigma Aldrich Supplier No. 7642025GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium trifluoromethanesulfonate is an aprotic neutral ionic liquid.1-Butyl-3-methylimidazolium trifluoromethanesulfonate can be used as an alternative to lithium perchlorate-diethyl ether mixture in the Diels-Alder reaction.

Specifications

CAS 174899-66-2
Density 1.292 g/mL at 20°C
Molecular Formula C9H15F3N2O3S
Refractive Index n20/D 1.434
Linear Formula C9H15F3N2O3S
MDL Number MFCD03427620
Quantity 5 g
Synonym BMIM Otf
Molecular Weight (g/mol) 288.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.