Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Trifluoromethanesulfonate

Catalog No. 764205GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-420-5GF 10 mg
76-420-25GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-420-5GF Supplier Sigma Aldrich Supplier No. 764205GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium trifluoromethanesulfonate is an aprotic neutral ionic liquid.1-Butyl-3-methylimidazolium trifluoromethanesulfonate can be used as an alternative to lithium perchlorate-diethyl ether mixture in the Diels-Alder reaction.

Specifications

Melting Point 62°C to 67°C
Molecular Formula C18H26N2O7
Linear Formula C18H26N2O7
Refractive Index n20/D 1.434
MDL Number MFCD22380740
Quantity 10 mg
Synonym BMIM Otf
Molecular Weight (g/mol) 382.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.