Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-4-ethynylbenzene 97.0+%, TCI America™
SDP

Catalog No. B23015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23015G 5 g
B230125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23015G Supplier TCI America Supplier No. B23015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79887-09-5
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00173885
InChI Key ZVWWYEHVIRMJIE-UHFFFAOYSA-N
Synonym 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene
PubChem CID 2775132
IUPAC Name 1-butyl-4-ethynylbenzene
SMILES CCCCC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 158.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-4-ethynylbenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.