Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-4-methylpyridinium Chloride 99.0+%, TCI America™
SDP

Catalog No. B34265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34265G 5 g
B342625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34265G Supplier TCI America Supplier No. B34265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112400-86-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD03095439
InChI Key YTSDTJNDMGOTFN-UHFFFAOYSA-M
PubChem CID 11074101
IUPAC Name 1-butyl-4-methylpyridin-1-ium chloride
SMILES [Cl-].CCCC[N+]1=CC=C(C)C=C1

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 185.70
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-4-methylpyridinium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.