Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-4-Methylpyridinium Tetrafluoroborate

Catalog No. 7326150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-261-50G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73-261-50G Supplier Sigma Aldrich Supplier No. 7326150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyridinium based ionic liquid4MBPBF4 has been used in the preparation of the SWNT-polymer composite films to improve the dispersion of SWNTs and conductivity of the composites (SWNT= singlewalled carbon nanotubes). The use of 4MBPBF4 as a reaction media during derivatization of dimethyl sulfate with dibenzazepine, accelerates the rate of the reaction. It can also be used to modify carbon paste electrode, which leads to high sensitivity, selectivity and low detection limit for both potassium ferricyanide and dopamine by cyclic voltammetric technique.Lipase catalyzed kinetic resolution in ionic liquids with improved enantioselectivity

Specifications

CAS 74839-84-2
Melting Point 242°C to 246°C
Molecular Formula C37H50Cl2N2ORu
Refractive Index n20/D 1.4
Linear Formula C37H50Cl2N2ORu
Quantity 2 g
Synonym Grubbs Catalyst(R) C711; Hoveyda-Grubbs Catalyst(R) M72 SIPr (C711); Dichloro[1,3-bis(2,6-isopropylphenyl)-2-imidazolidinylidene](2-isopropoxyphenylmethylene)ruthenium(II)
Molecular Weight (g/mol) 710.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.