Learn More
1-Butylamine, 99%
Description
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N'-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N′-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.
Solubility
Miscible with ethanol and water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, aluminum, copper, copper alloys and acids.
Specifications
Specifications
| Melting Point | -50°C |
| Density | 0.738 |
| Boiling Point | 76°C to 78°C |
| Flash Point | −14°C (6°F) |
| Odor | Ammonia-like |
| Refractive Index | 1.401 |
| Linear Formula | CH3CH2CH2CH2NH2 |
| Quantity | 2500 mL |
| UN Number | UN1125 |
| Beilstein | 605269 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H311+H331-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H302-H312-H332 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-BUTYLAMINE |
| EINECSNumber | 203-699-2 |
| RTECSNumber | EO2975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.