Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific™

Catalog No. AAH27661MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27661MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27661MD Supplier Thermo Scientific Chemicals Supplier No. H27661MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85167-10-8
Molecular Formula C14H25BO2
Molecular Weight (g/mol) 236.162
MDL Number MFCD09037499
InChI Key IMUJPBOUZJFWJB-FMSGJZPZSA-N
Synonym 3as,4s,6s,7ar-2-butyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole, 1s,2s,6r,8s-4-butyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0 2 ,? decane, 1s,2s,3r,5s-2,3-butylboranediylbisoxy pinane, 1-butylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, n-butylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,6r,8s-4-butyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-butyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 3as,4s,6s,7ar-2-butyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 1s,2s,6r,8s-4-butyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane
PubChem CID 10911528
SMILES B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCC

Specifications

Refractive Index 1.4629
Quantity 250 mg
Optical Rotation +25° (c=1 in Chloroform)
Formula Weight 236.17
Percent Purity 95%
Chemical Name or Material 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.