Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylboronic acid, 98%

Catalog No. AAA1372506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372506 5 g
AAA1372503 1 g
AAA1372514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1372506 Supplier Thermo Scientific Chemicals Supplier No. A1372506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4426-47-5 | C4H11BO2 | 101.94 g/mol

1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-47-5
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00002106
InChI Key QPKFVRWIISEVCW-UHFFFAOYSA-N
Synonym n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane
PubChem CID 20479
IUPAC Name butylboronic acid
SMILES CCCCB(O)O

Specifications

Melting Point 92°C to 96°C
Quantity 5 g
Beilstein 1733489
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
Formula Weight 101.94
Percent Purity 98%
Chemical Name or Material 1-Butylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-607-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.