Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butylimidazole

Catalog No. 34841425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-841-425G 25 g
34-841-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-841-425G Supplier Sigma Aldrich Supplier No. 34841425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-butylimidazole acts as an n-coordinated ligand
  • 1-butylimidazole is formed as intermediate during the synthesis of a cationic cyclodextrin, used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis
  • Influence of 1-butylimidazole as base on the synthesis of dissymmetric chlorohydrin ester has been reported.1-butylimidazole may be used in the synthesis of a heterocyclic mesomeric betaine
  • It was used in the synthesis of 1-(1-butyl-3-imidazolio)propane-3-sulfonate (bim3s)

Specifications

CAS 4316-42-1
Density 0.945 g/mL (at 25°C (literature))
Boiling Point 114°C to 116°C (12 mmHg)
Molecular Formula C7H12N2
Refractive Index n20/D 1.480 (literature)
Linear Formula C7H12N2
MDL Number MFCD00042753
Quantity 25 g
Molecular Weight (g/mol) 124.18
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.