Learn More
Medchemexpress LLC 1-Butynylbenzene-d5 | 1219803-38-9 | 135.22 g/mol | C10H5D5 | 50 MG
SDP
Catalog No. 5000283777
Encompass_Preferred
1-Butynylbenzene-d5 is a deuterium-labeled alkyne reagent intended for research use. It is designed for synthetic and analytical applications, including click chemistry and use as an internal standard in mass spectrometry or chromatography. The compound provides a stable isotope label to assist in tracing, quantification, and method development.
Catalog No. 50-002-83777 Supplier Medchemexpress LLC Supplier No. HYW142175S50MG
1-Butynylbenzene-d5 is a deuterium-labeled alkyne reagent intended for research use. It is designed for synthetic and analytical applications, including click chemistry and use as an internal standard in mass spectrometry or chromatography. The compound provides a stable isotope label to assist in tracing, quantification, and method development.