Learn More
Medchemexpress LLC 1-caffeoylquinic acid | 1241-87-8 | MFCD10566609 | 98.4% | 354.31 | C16H18O9 | 10 MG
SDP
Catalog No. 5000295863
Encompass_Preferred
1-Caffeoylquinic acid is a naturally occurring phenolic compound used as a research reagent with reported activity as an NF-κB inhibitor and a modulator of PD-1/PD-L1 interaction. It is supplied as a high-purity analytical standard for in vitro and analytical applications; handle and store according to supplier recommendations.
  • High purity (98.4%) suitable for analytical and biological assays.
  • Molecular formula C16H18O9 and molecular weight 354.31 for analytical reference.
  • Reported NF-κB inhibitory activity and modulation of PD-1/PD-L1 interaction.
  • Available in small research quantities, including 10 mg packs.
  • Store protected from light; solid storage at 4°C, in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-95863 Supplier Medchemexpress LLC Supplier No. HYN046010MG
1-Caffeoylquinic acid is a naturally occurring phenolic compound used as a research reagent with reported activity as an NF-κB inhibitor and a modulator of PD-1/PD-L1 interaction. It is supplied as a high-purity analytical standard for in vitro and analytical applications; handle and store according to supplier recommendations.
  • High purity (98.4%) suitable for analytical and biological assays.
  • Molecular formula C16H18O9 and molecular weight 354.31 for analytical reference.
  • Reported NF-κB inhibitory activity and modulation of PD-1/PD-L1 interaction.
  • Available in small research quantities, including 10 mg packs.
  • Store protected from light; solid storage at 4°C, in solvent -80°C (6 months) or -20°C (1 month).