Learn More
Medchemexpress LLC HY-N0460 1mg Medchemexpress, 1-Caffeoylquinic acid CAS:1241-87-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04601MG
Encompass_Preferred
Medchemexpress, HY-N0460 1mg 1-Caffeoylquinic acid CAS:1241-87-8 1-Caffeoylquinic acid is an effective NF-κB inhibitor, shows significant binding affinity to the RH domain of p105 with Ki of 0.002 μM and binding energy of 1.50 Kcal/mol. 1-Caffeoylquinic acid has anti-oxidative stress ability. 1-Caffeoylquinic acid inhibits PD-1/PD-L1 interact. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2791