Learn More
TARGETMOL CHEMICALS INC 1-Caffeoylquinic acid 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN10515MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1-Caffeoylquinic acid is an important intermediate in lignin biosynthesis. 1-Caffeoylquinic acid has anti-influenza, and antioxidant activities, it also slows the release of glucose into the bloodstream after a meal. Purity 99.16%
Catalog No. 50-287-9049