Learn More
Chemglass Life Sciences Cajon Cuffs Only, 1" Size, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF006803
Encompass_Preferred
Cajon Cuffs Only, 1" Size, Airfree, Schlenk. Cuff adapters MUST be used between AF-0067 unions and AF-0069 flexible tubing. They are made of #304 stainless steel, and the adapters do not require welding or brazing to tubing to insure a leak tight seal.
Catalog No. AF006803