Learn More
Chemglass Life Sciences Cajon Unions Only, Use with 1" OD Tube, Airfree, Schlenk
SDP
Catalog No. AF006706
Encompass_Preferred
Cajon Unions Only, Use with 1" OD Tube, Airfree, Schlenk. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel. O-Rings are 70 durometer Viton.
Catalog No. AF006706 Supplier Chemglass Life Sciences Supplier No. AF006706
Cajon Unions Only, Use with 1" OD Tube, Airfree, Schlenk. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel. O-Rings are 70 durometer Viton.